missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM1 Mouse anti-Human, Clone: 1G4, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59418-50UL
This item is not returnable.
View return policy
Description
PGAM1 Monoclonal specifically detects PGAM1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| PGAM1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1028, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| BPG-dependent PGAM 1, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, PGAMA, PGAM-Bphosphoglycerate mutase 1, phosphoglycerate mutase 1 (brain), phosphoglycerate mutase A, nonmuscle form, Phosphoglycerate mutase isozyme B | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1G4 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| PGAM1 | |
| Recombinant human PGAM1 (1-254aa) purified from E. coli | |
| 50 μL | |
| Stem Cell Markers | |
| 5223 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a λ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction