missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM1 Antibody (1G4), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59418R
This item is not returnable.
View return policy
Description
PGAM1 Monoclonal antibody specifically detects PGAM1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| PGAM1 | |
| Monoclonal | |
| DyLight 550 | |
| BPG-dependent PGAM 1, EC 3.1.3.13, EC 5.4.2.1, EC 5.4.2.4, PGAMA, PGAM-Bphosphoglycerate mutase 1, phosphoglycerate mutase 1 (brain), phosphoglycerate mutase A, nonmuscle form, Phosphoglycerate mutase isozyme B | |
| Recombinant human PGAM1 (1-254aa) purified from E. coli | |
| 0.1 mL | |
| Stem Cell Markers | |
| 5223 | |
| Store at 4°C in the dark. | |
| IgG2a λ |
| Western Blot, ELISA, Immunofluorescence | |
| 1G4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction