Learn More
Description
Pfu DNA Polymerase is a highly thermostable DNA polymerase from the hyperthermophilic archaeum Pyrococcus furiosus. The enzyme catalyzes the template-dependent polymerization of nucleotides into duplex DNA in the 5’→3’ direction. Pfu DNA Polymerase also exhibits 3’→5’ exonuclease (proofreading) activity that enables the polymerase to correct nucleotide incorporation errors. It has no 5’→3’ exonuclease activity.
Highlights
- Eight times more accurate than Taq DNA polymerase
- Highly thermostable – remains 95% active after 2 hours incubation at 95°C
- Generates blunt-end PCR products
- Incorporates modified nucleotides (e.g., biotin-, digoxigenin-, fluorescently-labeled nucleotides)
- Increased PCR yields with native Pfu DNA Polymerase
Specifications
Specifications
| Description | Pfu DNA Polymerase |
| Quantity | 500U |
| For Use With (Application) | High-fidelity PCR, Standard PCR, Cloning, RT-PCR, Site-directed Mutagenesis |
| Thermostability | remains 95% active after 2 hours incubation at 95°C |
| Includes | 2 x 1.25mL 10X Pfu Buffer with (NH4)2SO2, 2 x 1.25mL 10X Pfu Buffer with MgSO4 and 2 x 1.25mL 25mM MgSO4 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.