missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Pfu DNA Polymerase
useful for polymerization reactions requiring high-fidelity synthesis.
£213.00 - £824.00
Specifications
Content And Storage | Store at -20°C Guaranteed stable for 6 months when properly stored |
---|---|
For Use With (Application) | Standard PCR |
Concentration | 5 units/μL |
Storage Buffer | 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16002565
|
Abnova™
D0014.500UNITS |
500 units |
£213.00
500 units |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082555
|
Abnova™
D0014.1000UNITS |
1000 units |
£414.00
1000 units |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16092555
|
Abnova™
D0014.2500UNITS |
2500 units |
£824.00
2500 units |
Estimated Shipment: 11-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- In addition to 5′-3′ DNA polymerase activity, it also possesses 3′-5′ exonuclease (proofreading) activity.
- Pfu DNA Polymerase exhibits the lowest error rate of any thermostable DNA polymerase studied, is even up to ten fold more accurate than normal Taq DNA polymerase.
- Consequently, Pfu DNA Polymerase is useful for polymerization reactions requiring high-fidelity synthesis.
Specifications
Store at -20°C Guaranteed stable for 6 months when properly stored | |
Standard PCR | |
5 units/μL | |
20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol |