missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Pfu DNA Polymerase

Product Code. 16002565
Click to view available options
Quantity:
1000 units
2500 units
500 units
Unit Size:
1000 units
2500 units
500 units
This item is not returnable. View return policy

Product Code. 16002565

Brand: Abnova™ D0014.500Units

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

useful for polymerization reactions requiring high-fidelity synthesis.

  • In addition to 5′-3′ DNA polymerase activity, it also possesses 3′-5′ exonuclease (proofreading) activity.
  • Pfu DNA Polymerase exhibits the lowest error rate of any thermostable DNA polymerase studied, is even up to ten fold more accurate than normal Taq DNA polymerase.
  • Consequently, Pfu DNA Polymerase is useful for polymerization reactions requiring high-fidelity synthesis.

Specifications

Concentration 5 U/μL
Content And Storage Store at -20°C Guaranteed stable for 6 months when properly stored
Quantity 500 units
For Use With (Application) Standard PCR
Storage Buffer 20 mM Tris-HCl (pH 8.0), 1 mM DTT, 0.1 mM EDTA, 100 mM KCl, 0.5% Nonidet P40, 0.5% Tween 20 and 50% glycerol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.