missing translation for 'onlineSavingsMsg'
Learn More

VACUUBRAND™ PFPE Oil

Product Code. 10554902 Shop All Vacuubrand Products
Change view
Click to view available options
Volume (Metric):
0.5 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
10554902 0.5 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10554902 Supplier VACUUBRAND™ Supplier No. 20687600

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This synthetic oil has excellent chemical resistance

This synthetic oil has excellent chemical resistance. Therefore it is often used for pumping strong oxidants (halo-genides, nitrogen oxides, etc.). As this oil type must not be mixed with mineral oils, a pump intended for this oil should be built with it from the outset. For changeover of an existing pump to this oil the pump has to be completely disassembled, cleaned and refilled with perfluoropolyether oil at VACUUBRAND;PFPE oil type I for VACUUBRAND rotary vane pumps RE 2.5, RZ 2.5, RE 6, RZ 6;PFPE oil type II for all VACUUBRAND rotary vane pumps.
  • Low vapor pressure even at high temperatures
  • excellent lubricating properties
  • low oil backflow
  • excellent resistance to aging
TRUSTED_SUSTAINABILITY

Specifications

Product Type Oil 25/5
For Use With (Application) Used for pumping strong oxidants (Halogenides, Nitrogen Oxides, etc.)
Material PFPE
Packaging Bottle
Pressure <3 x 10-5mbar
Quantity 1 Ea.
Viscosity 60 mm2/sec.
Volume (Metric) 0.5 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.