missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PFP-BETA1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17919772
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17919772 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17919772 Supplier Invitrogen™ Supplier No. PA5144382

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PFP-BETA1 Polyclonal Antibody for Western Blot, ELISA

Catalytic subunit of pyrophosphate--fructose 6-phosphate 1-phosphotransferase. Catalyzes the phosphorylation of D-fructose 6-phosphate, the first committing step of glycolysis. Uses inorganic phosphate (PPi) as phosphoryl donor instead of ATP like common ATP-dependent phosphofructokinases (ATP-PFKs), which renders the reaction reversible, and can thus function both in glycolysis and gluconeogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PFP-BETA1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene PFP-BETA1
Gene Accession No. Q8W4M5
Gene Alias PFP-BETA1 At1g12000 F12F1.13; Pyrophosphate-fructose 6-phosphate 1-phosphotransferase subunit beta 1
Gene Symbols PFP-BETA1
Host Species Rabbit
Immunogen Recombinant Arabidopsis thaliana PFP-BETA1 protein (157-427aa ).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 837752
Target Species Plant
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.