missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFM 01
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PFM 01 is a MRE11 endonuclease inhibitor. Prevents double-stranded break (DSB) resection. Enhances non-homologous end-joining (NHEJ) and reduces homologous recombination.
Specifications
Specifications
| Chemical Name or Material | (5Z)-5-[(4-Hydroxyphenyl)methylene]-3-(2-methylpropyl)-2-thioxo-4-thiazolidinone |
| CAS | 1558598-41-6 |
| Assay Percent Range | >98% |
| Quantity | 50 mg |
| Molecular Formula | C14H15NO2S2 |
| Formula Weight | Observed MW: 293.4 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction