missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PFKP Monoclonal antibody specifically detects PFKP in Human, Mouse, Rat samples. It is validated for Flow Cytometry
Specifications
Specifications
| Antigen | PFKP |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | OTI1D6 |
| Conjugate | mFluor Violet 500 SE |
| Dilution | Flow Cytometry |
| Formulation | 50mM Sodium Borate |
| Gene Alias | 6-phosphofructokinase type C, 6-phosphofructokinase, platelet type, EC 2.7.1, EC 2.7.1.11, PFK-CFLJ40226, PFKFplatelet type, Phosphofructokinase 1, phosphofructokinase, platelet, Phosphohexokinase |
| Host Species | Mouse |
| Immunogen | Full length human recombinant protein of human PFKP(NP_002618) produced in HEK293T cell. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?