missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKP Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-37947-100ul
This item is not returnable.
View return policy
Description
PFKP Polyclonal antibody specifically detects PFKP in Human samples. It is validated for ELISA,Western Blot
Specifications
| PFKP | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| 6-phosphofructokinase type C, 6-phosphofructokinase, platelet type, EC 2.7.1, EC 2.7.1.11, PFK-CFLJ40226, PFKFplatelet type, Phosphofructokinase 1, phosphofructokinase, platelet, Phosphohexokinase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 605-784 of human PFKP (NP_002618.1).,, Sequence:, IFEEPFDIRDLQSNVEHLTEKMKTTIQRGLVLRNESCSENYTTDFIYQLYSEEGKGVFDCRKNVLGHMQQGGAPSPFDRNFGTKISARAMEWITAKLKEARGRGKKFTTDDSICVLGISKRNVIFQPVAELKKQTDFEHRIPKEQWWLKLRPLMKILAKYKASYDVSDSGQLEHVQPWSV | |
| 100 μL | |
| Lipid and Metabolism | |
| 5214 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction