missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PFKL Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA521685

Product Code. 13249058

  • £481.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: A431, H1299, HeLa, HepG2. Predicted reactivity: Mouse (95%), Rat (95%), Chicken (88%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Phosphofructokinase (PFK), a major regulatory enzyme in all cells of the body, catalyzes the metabolism of sugar, and thereby is pivotal in the production of energy to maintain normal cell function. In human there are three structural loci controlling PFK: M (muscle), L (liver), and P (platelet) type subunits, which are variably expressed in different tissues; human diploid fibroblasts and leukocytes express all three genes. PFK, a tetramer formed by the random association of the products of two separate gene loci to form the five possible tetramers. PFKs of muscle and liver are homotetramers of the M and L subunits, respectively. Red cells have all five isozymes: M4, M3L, M2L2, ML3, and L4. PFK is an allosteric enzyme activated by ADP, AMP, or fructose bisphosphate and inhibited by ATP or citrate. PFK catalyzes the key controlling step of glycolytic pathway. PFK deficiency can present as mild to life-threatening episodic illness. A hallmark sign of this disease is intermittent dark urine, with the color of the urine ranging from orange to dark coffee-brown, which commonly develops following strenuous exercise. The mean red cell PFK is elevated in persons with Down syndrome.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PFKL
Polyclonal
Unconjugated
PFKL
6-phosphofructokinase type B; 6-phosphofructokinase, liver type; AA407869; ATP-dependent 6-phosphofructokinase, liver type; ATP-dependent 6-phosphofructokinase, liver type {ECO:0000255; ATP-PFK; ATP-PFK {ECO:0000255; EC 2.7.1.11; HAMAP-Rule:MF_03184}; K6PL; liver-type 1-phosphofructokinase; Pfkb; PFK-B; Pfkl; Pfk-l; Phosphofructo-1-kinase isozyme B; phosphofructokinase 1; phosphofructokinase, liver; phosphofructokinase, liver type; phosphofructokinase, liver, B-type; phosphohexokinase; phosphohexokinase {ECO:0000255
Rabbit
Antigen affinity chromatography
RUO
18641, 5211
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1.23 mg/mL
PBS with 20% glycerol and 0.025% ProClin 300; pH 7
P12382, P17858
PFKL
Recombinant fragment corresponding to a region within amino acids 428 and 674 of PFKL (Uniprot ID#P17858).
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.