missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKL Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-32212CL1
This item is not returnable.
View return policy
Description
PFKL Polyclonal antibody specifically detects PFKL in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| PFKL | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| 6-phosphofructokinase, liver type, DKFZp686G1648, DKFZp686L2097, EC 2.7.1, EC 2.7.1.11, FLJ30173, FLJ40909, human liver-type 1-phosphofructokinase, EC 2.7.1.1110Phosphohexokinase, liver-type 1-phosphofructokinase, PFK-B, Phosphofructo-1-kinase isozyme B, Phosphofructokinase 1, phosphofructokinase, liver, phosphohexokinase | |
| The immunogen this antibody was made to, maps to a region between residue 1 to 50 of human Phosphofructokinase, Liver using the numbering given in entry NP_002617.3 (GeneID 5211). | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 5211 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction