missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PFKFB4 Recombinant Rabbit Monoclonal Antibody (HL2204)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA550430
This item is not returnable.
View return policy
Description
PFKFB4 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Synthesis and degradation of fructose 2,6-bisphosphate.
Specifications
| PFKFB4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q16877 | |
| PFKFB4 | |
| Recombinant fragment of human PFKFB4. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| HL2204 | |
| Unconjugated | |
| PFKFB4 | |
| 6-biphosphatase-4; 6PF-2-K/Fru-2,6-P2ase 4; 6PF-2-K/Fru-2,6-P2ASE testis-type isozyme; 6-phosphofructo-2-kinase; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4; 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4; 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 4; bifunctional enzyme with kinase and biphosphatase activities; C230090D14; Fructose-2,6-bisphosphatase; PFK/FBPase 4; Pfkfb4 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 5210 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction