missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKFB4 Antibody (OTI1C8), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73340APC
This item is not returnable.
View return policy
Description
PFKFB4 Monoclonal antibody specifically detects PFKFB4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| PFKFB4 | |
| Monoclonal | |
| APC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 5210 | |
| Store at 4C in the dark. | |
| IgG2a |
| Western Blot, Immunocytochemistry | |
| OTI1C8 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 4,6-phosphofructo-2-kinase/fructose-2,6-biphosphatase-4,6PF-2-K/Fru-2,6-P2ase 4,6PF-2-K/Fru-2,6-P2ase testis-type isozyme, bifunctional enzyme with kinase and biphosphatase activities, PFK/FBPase 4 | |
| Full length human recombinant protein of human PFKFB4 (NP_004558) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction