missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFKFB2 [p Ser483] Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38640-100ul
This item is not returnable.
View return policy
Description
PFKFB2 Polyclonal antibody specifically detects PFKFB2 in Human samples. It is validated for ELISA,Western Blot
Specifications
| PFKFB2 | |
| Polyclonal | |
| Western Blot 1:1000 - 1:5000, ELISA | |
| 6PF-2-K/Fru-2,6-P2ase 2, 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2, 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase 2, DKFZp781D2217, fructose-2,6-bisphosphatase, cardiac isozyme, MGC138308,6PF-2-K/Fru-2,6-P2ase heart-type isozyme, MGC138310, PFK/FBPase 2, PFK-2/FBPase-2, PFKFB, cardiac | |
| A synthetic phosphorylated peptide around S483 of human PFKFB2 (NP_006203.2).,, Sequence:, NYSVG | |
| 100 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 5208 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction