missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PFI 7

Product Code. p-200112509
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30279878

Brand: Tocris Bioscience™ 7960/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective GID4 antagonist

PFI 7 is a selective GID4 antagonist (Kd = 80 nM by SPR assays and EC50 = 0.6 μM by cell-based NanoBRET). PFI 7 binds within the ?-barrel of GID4 substrate binding pocket, disrupting its interaction with the canonical Pro/N-degron peptide in live cells. It alters protein levels of several proteins including RNA helicases as measured by quantitative proteomics.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-((1s,4s)-4-(1H-Benzo[d]imidazol-2-yl)cyclohexyl)-2-(((1H-indol-2-yl)methyl)amino)acetamide dihydrochloride
Quantity 50 mg
Target Ubiquitin Ligase (E3) Inhibitors
Molecular Formula C24H27N5O.2HCl
Purity 0.98
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.