missing translation for 'onlineSavingsMsg'
Learn More

PFI-2 hydrochloride, MedChemExpress

Product Code. p-200100547
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16453567

Brand: Medchem Express HY18627A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PFI-2 ((R)-PFI-2 hydrochloride) hydrochloride is a potent and selective SET domain containing lysine methyltransferase 7 (SETD7) inhibitor. (R)-PFI-2 shows high inhibiting activity with IC50 value of 2.0 nM and (S)-PFI-2 shows inhibiting activity with IC50 value of 1.0 μM. PFI-2 hydrochloride can be used for the research of chronic kidney disease and inflammation response in the development of renal fibrosis.

PFI-2 ((R)-PFI-2 hydrochloride) hydrochloride is a potent and selective SET domain containing lysine methyltransferase 7 (SETD7) inhibitor. (R)-PFI-2 shows high inhibiting activity with IC50 value of 2.0 nM and (S)-PFI-2 shows inhibiting activity with IC50 value of 1.0 μM. PFI-2 hydrochloride can be used for the research of chronic kidney disease and inflammation response in the development of renal fibrosis.

Specifications

Chemical Name or Material PFI-2 hydrochloride
CAS 1627607-87-7
Color White
Purity Grade Notes Research
Molecular Formula C23H26ClF4N3O3S
Quantity 50 mg
Synonym (R)-PFI-2 hydrochloride
Solubility Information DMSO : ≥ 32 mg/mL (59.70 mM) ∣H2O : 5.56 mg/mL (10.37 mM; Need ultrasonic)
SMILES O=S(C1=CC2=C(CNCC2)C(F)=C1)(N[C@H](CC3=CC=CC(C(F)(F)F)=C3)C(N4CCCC4)=O)=O.[H]Cl
Molecular Weight (g/mol) 535.98
Formula Weight 535.98
Percent Purity 96.09%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.