missing translation for 'onlineSavingsMsg'
Learn More

PfHsp70 Rabbit anti-P. falciparum, Polyclonal, Novus Biologicals™

Product Code. 18300847 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18300847 25 μg 25µg
18320535 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18300847 Supplier Novus Biologicals Supplier No. NBP31823925UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PfHsp70 Polyclonal antibody specifically detects PfHsp70 in P. falciparum samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PfHsp70
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:50
Formulation PBS pH 7.4, 50% glycerol
Gene Alias Heat shock 70 kDa protein
Host Species Rabbit
Immunogen His-tagged and purified PfHSP70, C-terminus (AA 365-681)
Purification Method Protein A purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species P. falciparum
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.