missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PFDN6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA587996
This item is not returnable.
View return policy
Description
Immunogen sequence: MAELIQKKLQ GEVEKYQQLQ KDLSKSMSGR QKLEAQLTEN NIVKEELALL DGSNVVFKLL GPVLVKQELG EARATVGKRL DYITAEIKRY ESQLRDLERQ SEQQRETLAQ LQQEFQRAQA AKAGAPGKA; Positive Samples: Mouse liver.
PFDN6 binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. This protein also binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.
Specifications
| PFDN6 | |
| Polyclonal | |
| Unconjugated | |
| Pfdn6 | |
| H2-K region expressed gene 2; H-2Ke2; H2-KE2; HKE2; HLA class II region expressed gene KE2; hypothetical protein LOC393485; KE2; KE-2; KE2 protein; MHC class II region expressed gene KE2; Pfd6; PFDN6; prefoldin 6; prefoldin subunit 6; Protein Ke2; zgc:66282 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14976 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 4.43 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q03958 | |
| Pfdn6 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-129 of human PFDN6 (NP_0550751). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction