missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN6 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£155.00 - £364.00
Specifications
| Antigen | PFDN6 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
PFDN6 Polyclonal antibody specifically detects PFDN6 in Mouse samples. It is validated for Western BlotSpecifications
| PFDN6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 10471 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse | |
| H2-KE2, HLA class II region expressed gene KE2, KE-2, KE2 protein, MGC70744, PFD6HKE2, prefoldin 6, prefoldin subunit 6, Protein Ke2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-129 of human PFDN6 (NP_055075.1). MAELIQKKLQGEVEKYQQLQKDLSKSMSGRQKLEAQLTENNIVKEELALLDGSNVVFKLLGPVLVKQELGEARATVGKRLDYITAEIKRYESQLRDLERQSEQQRETLAQLQQEFQRAQAAKAGAPGKA | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title