Learn More
Invitrogen™ PFDN5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521380
Description
Recommended positive controls: 293T, A431, HepG2. Predicted reactivity: Mouse (100%), Rat (99%), Zebrafish (88%), Xenopus laevis (85%), Dog (98%), Rhesus Monkey (100%), Bovine (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins. Represses the transcriptional activity of MYC.
Specifications
| PFDN5 | |
| Polyclonal | |
| Unconjugated | |
| PFDN5 | |
| 1190001O17Rik; 1700010A06Rik; c-myc binding protein; c-myc binding protein MM-1; C-myc-binding protein Mm-1; D15Ertd697e; Eig1; EIG-1; MM1; MM-1; Myc modulator 1; myc modulator-1; PFD5; Pfdn5; prefoldin 5; prefoldin subunit 5 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5204 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.56 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q99471 | |
| PFDN5 | |
| Recombinant protein encompassing a sequence within the center region of human Prefoldin 5. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.