missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN4 Antibody (02), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06528PCP
This item is not returnable.
View return policy
Description
PFDN4 Monoclonal antibody specifically detects PFDN4 in Human samples. It is validated for ELISA
Specifications
| PFDN4 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 02 | |
| ELISA | |
| C-1, PFD4C1prefoldin 4, prefoldin subunit 4, Protein C-1 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PFDN4 (Uniprot#: Q9NQP4; Met1-Ser134). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 5203 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction