missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13053-1-AP-150UL
This item is not returnable.
View return policy
Description
Binds specifically to cytosolic chaperonin (c-CPN) and transfers target proteins to it. Binds to nascent polypeptide chain and promotes folding in an environment in which there are many competing pathways for nonnative proteins.Specifications
| PFDN2 | |
| Polyclonal | |
| Unconjugated | |
| PFDN2 | |
| PFD2, PFDN2, prefoldin subunit 2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18637, 5202 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O70591, Q9UHV9 | |
| Pfdn2 | |
| PFDN2 Fusion Protein Ag3720 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction