missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PFDN2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£178.00 - £255.00
Specifications
| Antigen | PFDN2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate |
| Applications | Western Blot, ELISA, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18613437
|
Novus Biologicals
NBP3-00279-100ul |
100 μg |
£255.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18646238
|
Novus Biologicals
NBP3-00279-50ul |
50 μg |
£178.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PFDN2 Polyclonal antibody specifically detects PFDN2 in Human samples. It is validated for Western Blot, ELISA, ImmunoprecipitationSpecifications
| PFDN2 | |
| Western Blot, ELISA, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 5202 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, ELISA 1:5000-1:10000, Immunoprecipitation 1-4 uL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| PFD2, prefoldin 2, prefoldin subunit 2 | |
| Produced in rabbits immunized with purified, recombinant Human PFDN2 (Uniprot#: Q9UHV9; Met1-Ser154) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title