missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRANDTECH™ VITLAB™ PFA Wide-Mouth Reagent Bottles
Click to view available options
Capacity (Metric):
2000 mL
Unit Size:
Each
Description
- Leach-resistant bottles for long-term storage of standards
- Wide mouth for easier access to contents
- PFA screw caps help protect against outside contamination and leakage
Specifications
Specifications
| Capacity | 64 oz., 2000mL |
| Capacity (Metric) | 2000 mL |
| Capacity (English) | 64 oz. |
| Color | Translucent |
| Diameter (English) | 5.1 in. |
| Material | PFA |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide, Threaded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction