Learn More
Cole-Parmer™ PFA Threaded Plug
Product Code. 11775758
Click to view available options
Thread Size:
1/2 in.
3/8 in.
Unit Size:
Each
Product Code. 11775758
Brand: Cole-Parmer™ 0654640
Only null left
Add to basket
Add to basket
Threaded fittings offer more permanent installation
  • Chemically-inert PFA handles most aggressive chemicals
  • Threaded reducing bushing
  • Choose the size and material that meets your application needs
  • Use with our rigid tubing for your high-pressure applications
Specifications
Type Plug
Color Clear
Thread Size 1/2 in.
Port Type Male NPT Fitting
Sterilization Method Ethylene Oxide (ETO) Only
Overall Length (English) 27/32 in.
Material PFA
Overall Width (English) 15/16 in.