missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ PFA Reagent Bottle

Product Code. 10283452 Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
1000 mL
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10283452

Brand: BRAND™ 128962

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Designed for long-term storage of solvents and standards (trace analytics). PFA screw cap with a molded sealing ring provides a positive, leak-resistant seal.

  • PerFluoroAlkoxy (PFA) construction
  • Narrow neck
  • Autoclavable
  • Easy to clean
  • Withstands temperatures from -200 to 260°C
  • Includes thread S28 and screw cap
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Color Translucent
For Use With (Application) For long-term storage of solvents and standards
Fill Volume Range 1000mL
Includes Screw cap with a molded sealing ring for a positive seal
Material PFA
Closure Included Yes
Mouth Narrow
Height (Metric) 240 mm
Closure Material PFA
Closure Size S28
Diameter (Metric) 96 mm
Autoclavable Autoclavable
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.