missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ PFA Reagent Bottle
Shop All Brand Products
Click to view available options
Capacity (Metric):
50 mL
1000 mL
Unit Size:
Each
Description
- PerFluoroAlkoxy (PFA) construction
- Narrow neck
- Autoclavable
- Easy to clean
- Withstands temperatures from -200 to 260°C
- Includes thread S28 and screw cap
Specifications
Specifications
| Capacity (Metric) | 50 mL |
| Cleaning Procedure | Autoclaving |
| Color | Translucent |
| For Use With (Application) | Designed for long-term storage of solvents and standards |
| Material | PFA |
| Temperature Range (Metric) | –200 to 260°C |
| Closure Included | Yes |
| Mouth | Narrow |
| Product Type | Reagent Bottle |
| Closure Material | PFA |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction