missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ PFA Fitting

Product Code. 10671771 Shop All SCAT Europe Products
Change view
Click to view available options
Diameter (Metric) Outer:
1.6 mm
2.3 mm
3.2 mm
Unit Size:
Pack of 5
Product Code. Diameter (Metric) Outer unitSize
10671771 3.2 mm Pack of 5
10202090 1.6 mm Pack of 5
10292320 2.3 mm Pack of 5
3 options
This item is not returnable. View return policy

Product Code. 10671771

Brand: SCAT™ 107063

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.

SCAT™ fittings are available in the following versions:

  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters

Specifications

Type Fitting
Color Blue
Diameter (Metric) Outer 3.2 mm
Diameter (Metric) Inner 3.2 mm
For Use With (Application) Solvent Handling
Packaging Quantity 5 Fittings
For Use With Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems
Material PFA
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.