missing translation for 'onlineSavingsMsg'
Learn More

SCAT™ PFA Fitting

Product Code. 10202090 Shop All SCAT Europe Products
Change view
Click to view available options
Diameter (Metric) Outer:
1.6 mm
2.3 mm
3.2 mm
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer unitSize
10202090 1.6 mm Pack of 5
10292320 2.3 mm Pack of 5
10671771 3.2 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10202090 Supplier SCAT™ Supplier No. 107061
Only null left
Add to basket
Add to basket

Connect your HPLC capillaries safely and tightly to your SCAT™ system. SCAT™ PFA fittings are chemically resistant and fit Safety Caps and Safety Waste Caps. Available for different capillary sizes.

SCAT™ fittings are available in the following versions:

  • PFA fitting with integrated ferrule in various colors and diameters
  • Tube connectors in various diameters

Specifications

Type Fitting
Color Green
Diameter (Metric) Outer 1.6 mm
Diameter (Metric) Inner 1.6 mm
For Use With (Application) Solvent Handling
Packaging Quantity 5 Fittings
For Use With Safety Caps, Safety Waste Caps, HPLC and UHPLC Systems
Material PFA
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.