Learn More
Masterflex™ PFA Compression Threaded Adapter, Male NPT
Product Code. 12354538
Click to view available options
Thread Size:
1/4 in.
1/8 in.
Unit Size:
Each
Product Code. 12354538
Brand: Masterflex™ 3132163
Only null left
Add to basket
Add to basket
Superior Compression Threaded Adapter are chemical resistance with durable PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
Specifications
Description PFA compression threaded adapter, 4 mm tubing OD x 1/4 in. male NPT
Diameter (Metric) Outer 4 mm
Outlet Port Compression x NPT(M)
Sterilization Method Autoclave or ethylene oxide (ETO)
Thread Size 1/4 in.
Includes Compression Size / Tube Od (Port 1): 4 mm
Material (Seal) PFA
Product Type Vacuum Tubing