Learn More
PeproTech PF4 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Brand: PeproTech 500-P05BT-500UG
Description
AA Sequence of recombinant protein: EAEEDGDLQC LCVKTTSQVR PRHITSLEVI KAGPHCPTAQ LIATLKNGRK ICLDLQAPLY KKIIKKLLES. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human PF-4 (CXCL4). Anti-Human PF-4 (CXCL4)-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect Human PF-4 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Human PF-4 (500-P05) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human PF-4. Western Blot: To detect hPF-4 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hPF-4 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P05BT-1MG will be provided as 2 x 500 μg
Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).
Specifications
| PF4 | |
| Polyclonal | |
| Biotin | |
| Pf4 | |
| chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5196 | |
| -20°C | |
| Lyophilized |
| ELISA, Western Blot | |
| 0.1-1.0 mg/mL | |
| PBS with no preservative | |
| P02776 | |
| Pf4 | |
| E.coli-derived, 7.8 kDa Recombinant Human PF-4 (CXCL4). | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.