missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PF4 Monoclonal Antibody (RTO)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA517639
This item is not returnable.
View return policy
Description
MA5-17639 targets Platelet Factor 4 in ELISA and WB applications and shows reactivity with Human samples. The MA5-17639 immunogen is human Factor IV purified from the supernatant of thrombin activated platelets.
Platelet factor-4 is a 70-amino acid protein that is released from the alpha-granules of activated platelets and binds with high affinity to heparin. Its major physiologic role appears to be neutralization of heparin-like molecules on the endothelial surface of blood vessels, thereby inhibiting local antithrombin III activity and promoting coagulation. As a strong chemoattractant for neutrophils and fibroblasts, PF4 probably has a role in inflammation and wound repair (Eisman et al., 1990).
Specifications
| PF4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P02776 | |
| Pf4 | |
| Human Factor IV purified from the supernatant of thrombin activated platelets. | |
| 200 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b κ |
| ELISA, Western Blot | |
| RTO | |
| Unconjugated | |
| Pf4 | |
| chemokine (C-X-C motif) ligand 4; C-X-C motif chemokine 4; Cxcl4; Endothelial cell growth inhibitor; Iroplact; oncostatin-A; PF4; PF-4; Pf4a; Platelet factor 4; Platelet factor 4, short form; RATPF4A; Scyb4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5196 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction