missing translation for 'onlineSavingsMsg'
Learn More

PF-CBP1 hydrochloride, MedChemExpress

Product Code. 16488008
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488008 10 mg 10mg
16467786 100 mg 100mg
16433937 50 mg 50mg
16433736 25 mg 25mg
16420478 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16488008 Supplier Medchem Express Supplier No. HY19999A

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress PF-CBP1 hydrochloride is a highly selective inhibitor of the CREB binding protein bromodomain (CBP BRD).

PF-CBP1 inhibits CREBBP and EP300 bromodomains with IC50 of 125 nM and 363 nM respectively. PF-CBP1 hydrochloride reduces LPS-induced inflammatory cytokines expression (IL-1β, IL-6 and IFN-β) in primary macrophages. PF-CBP1 hydrochloride also downregulates RGS4 expression cortical neurons and can be used for the research of neurological disorders, including epilepsy and parkinson's disease, et al.

Specifications

Chemical Name or Material PF-CBP1 hydrochloride
CAS 2070014-93-4
Color White
Purity Grade Notes Research
Molecular Formula C29H37ClN4O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (190.45 mM; Need ultrasonic) ∣H2O : 1 mg/mL (1.90 mM; Need ultrasonic)
SMILES CCCOC1=CC=C(CCC2=NC3=CC(C4=C(C)ON=C4C)=CC=C3N2CCN5CCOCC5)C=C1.Cl[H]
Molecular Weight (g/mol) 525.08
Formula Weight 525.08
Percent Purity 95.95%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.