missing translation for 'onlineSavingsMsg'
Learn More

PF-8380, MedChemExpress

Product Code. 16448357
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16448357

Brand: Medchem Express HY13344

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-8380 is a potent autotaxin inhibitor with an IC50 of 2.8 nM in isolated enzyme assay and 101 nM in human whole blood.

PF-8380 is a potent autotaxin inhibitor with an IC50 of 2.8 nM in isolated enzyme assay and 101 nM in human whole blood.

Specifications

Chemical Name or Material PF-8380
CAS 1144035-53-9
Color White
Purity Grade Notes Research
Molecular Formula C22H21Cl2N3O5
Quantity 5 mg
Solubility Information DMSO : 6.67 mg/mL (13.94 mM; ultrasonic and adjust pH to 5 with HCl)
SMILES O=C(N1CCN(CC1)CCC(C2=CC(O3)=C(C=C2)NC3=O)=O)OCC4=CC(Cl)=CC(Cl)=C4
Molecular Weight (g/mol) 478.33
Formula Weight 478.33
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.