missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-670462, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PF-670462 is a potent and selective inhibitor of casein kinase (CK1ε and CK1δ), with IC50s of 7.7 nM and 14 nM, respectively.
Specifications
Specifications
| Chemical Name or Material | PF-670462 |
| CAS | 950912-80-8 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C19H22Cl2FN5 |
| Quantity | 50 mg |
| Solubility Information | H2O : 100 mg/mL (243.71 mM; Need ultrasonic) ∣DMSO : ≥ 32 mg/mL (77.99 mM) |
| SMILES | NC1=NC=CC(C2=C(C3=CC=C(F)C=C3)N=CN2C4CCCCC4)=N1.[H]Cl.[H]Cl |
| Molecular Weight (g/mol) | 410.32 |
| Formula Weight | 410.32 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction