missing translation for 'onlineSavingsMsg'
Learn More

PF-670462, MedChemExpress

Product Code. 16416578
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 16416578

Brand: Medchem Express HY15490

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-670462 is a potent and selective inhibitor of casein kinase (CK1ε and CK1δ), with IC50s of 7.7 nM and 14 nM, respectively.

PF-670462 is a potent and selective inhibitor of casein kinase (CK1ε and CK1δ), with IC50s of 7.7 nM and 14 nM, respectively.

Specifications

Chemical Name or Material PF-670462
CAS 950912-80-8
Color White
Purity Grade Notes Research
Molecular Formula C19H22Cl2FN5
Quantity 10 mg
Solubility Information H2O : 100 mg/mL (243.71 mM; Need ultrasonic) ∣DMSO : ≥ 32 mg/mL (77.99 mM)
SMILES NC1=NC=CC(C2=C(C3=CC=C(F)C=C3)N=CN2C4CCCCC4)=N1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 410.32
Formula Weight 410.32
Percent Purity 98.49%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.