missing translation for 'onlineSavingsMsg'
Learn More

PF-543 Citrate, MedChemExpress

Product Code. 16442607
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16442607

Brand: Medchem Express HY15425A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-543 Citrate (Sphingosine Kinase 1 Inhibitor II Citrate) is a potent, selective, reversible and sphingosine-competitive SPHK1 inhibitor with an IC50 of 2 nM and a Ki of 3.6 nM. PF-543 Citrate is >100-fold selectivity for SPHK1 over SPHK2. PF-543 Citrate is an effective potent inhibitor of sphingosine 1-phosphate (S1P) formation in whole blood with an IC50 of 26.7 nM. PF-543 Citrate induces apoptosis, necrosis, and autophagy.

PF-543 Citrate (Sphingosine Kinase 1 Inhibitor II Citrate) is a potent, selective, reversible and sphingosine-competitive SPHK1 inhibitor with an IC50 of 2 nM and a Ki of 3.6 nM. PF-543 Citrate is >100-fold selectivity for SPHK1 over SPHK2. PF-543 Citrate is an effective potent inhibitor of sphingosine 1-phosphate (S1P) formation in whole blood with an IC50 of 26.7 nM. PF-543 Citrate induces apoptosis, necrosis, and autophagy.

Specifications

Chemical Name or Material PF-543 Citrate
CAS 1415562-83-2
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C33H39NO11S
Quantity 50 mg
Synonym Sphingosine Kinase 1 Inhibitor II Citrate
Solubility Information DMSO : ≥ 100 mg/mL (152.04 mM) ∣H2O : 50 mg/mL (76.02 mM; Need ultrasonic)
SMILES OC(C(O)=O)(CC(O)=O)CC(O)=O.O=S(C1=CC=CC=C1)(CC2=CC(C)=CC(OCC3=CC=C(CN4[C@@H](CO)CCC4)C=C3)=C2)=O
Molecular Weight (g/mol) 657.73
Formula Weight 657.73
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.