missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-543 Citrate, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
PF-543 Citrate (Sphingosine Kinase 1 Inhibitor II Citrate) is a potent, selective, reversible and sphingosine-competitive SPHK1 inhibitor with an IC50 of 2 nM and a Ki of 3.6 nM. PF-543 Citrate is >100-fold selectivity for SPHK1 over SPHK2. PF-543 Citrate is an effective potent inhibitor of sphingosine 1-phosphate (S1P) formation in whole blood with an IC50 of 26.7 nM. PF-543 Citrate induces apoptosis, necrosis, and autophagy.
Specifications
Specifications
| Chemical Name or Material | PF-543 Citrate |
| CAS | 1415562-83-2 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C33H39NO11S |
| Quantity | 5 mg |
| Synonym | Sphingosine Kinase 1 Inhibitor II Citrate |
| Solubility Information | DMSO : ≥ 100 mg/mL (152.04 mM) ∣H2O : 50 mg/mL (76.02 mM; Need ultrasonic) |
| SMILES | OC(C(O)=O)(CC(O)=O)CC(O)=O.O=S(C1=CC=CC=C1)(CC2=CC(C)=CC(OCC3=CC=C(CN4[C@@H](CO)CCC4)C=C3)=C2)=O |
| Molecular Weight (g/mol) | 657.73 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction