missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 5006739

Product Code. 18770959
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18770959 25 mg 25mg
18712458 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18770959 Supplier Tocris Bioscience™ Supplier No. 6346/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent CK1δ/ε inhibitor

PF 5006739 is a potent casein kinase 1δ/ε inhibitor (IC50 values are 3.9 and 17 nM, respectively). Mediates circadian rhythm phase-delaying effects in vivo. Attenuates opioid drug seeking behavior in addiction in vivo models.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-[4-(4-Fluorophenyl)-1-[1-(3-isoxazolylmethyl)-4-piperidinyl]-1H-imidazol-5-yl]-2-pyrimidinamine
CAS 1293395-67-1
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C22H22FN7O
Formula Weight Observed MW: 419.46
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.