missing translation for 'onlineSavingsMsg'
Learn More

PF-4989216, MedChemExpress

Product Code. 16495888
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16495888 10 mg 10mg
16492037 50 mg 50mg
16486346 100 mg 100mg
16418537 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16495888 Supplier Medchem Express Supplier No. HY13864

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-4989216 is a potent and selective PI3Kα inhibitor with a Ki of 0.6 nM.

PF-4989216 is a potent and selective PI3Kα inhibitor with a Ki of 0.6 nM.

Specifications

Chemical Name or Material PF-4989216
CAS 1276553-09-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H13FN6OS
Quantity 10 mg
Solubility Information DMSO : ≥ 29 mg/mL (76.24 mM)
SMILES N#CC1=C(N2CCOCC2)SC(C3=NC=NN3)=C1C4=CC=C(C#N)C=C4F
Molecular Weight (g/mol) 380.4
Formula Weight 380.4
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.