missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 4800567 hydrochloride
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Selective casein kinase 1ε inhibitor; displays 22-fold greater potency towards CK1ε than CK1δ (IC50 values are 32 and 711 nM for CK1ε and CK1δ respectively). ATP competitive. Displays minimal effect on the circadian clock.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 396.27 |
| Purity | >99% |
| Molecular Weight (g/mol) | 396.272 |
| Chemical Name or Material | PF 4800567 hydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction