missing translation for 'onlineSavingsMsg'
Learn More

PF-4191834, MedChemExpress

Product Code. 16437617
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437617 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16437617 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY117048

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-4191834 (PF-04191834) is an orally active, noniron chelating, and non-redox inhibitor of the 5-Lipoxygenase (5-LOX) (IC50=229 nM), displays ∼300-fold selectivity for 5-LOX over 12-LOX and 15-LOX, shows no activity toward the cyclooxygenase enzymes, and is effective in inflammation and pain.

PF-4191834 (PF-04191834) is an orally active, noniron chelating, and non-redox inhibitor of the 5-Lipoxygenase (5-LOX) (IC50=229 nM), displays ∼300-fold selectivity for 5-LOX over 12-LOX and 15-LOX, shows no activity toward the cyclooxygenase enzymes, and is effective in inflammation and pain.

Specifications

Chemical Name or Material PF-4191834
Physical Form Solid
CAS 1029317-21-2
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C22H23N3O2S
For Use With (Application) Neuroscience-Neuromodulation
Synonym PF-04191834
Solubility Information DMSO : 100 mg/mL (254.13 mM; Need ultrasonic)
SMILES O=C(C1(C2=CC=CC(SC3=CC=C(C4=CC=NN4C)C=C3)=C2)CCOCC1)N
Molecular Weight (g/mol) 393.5
Formula Weight 393.5
Percent Purity 98.62%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.