missing translation for 'onlineSavingsMsg'
Learn More

PF-3845, MedChemExpress

Product Code. 16456048
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16456048 10 mg 10mg
16452187 50 mg 50mg
16418657 5 mg 5mg
16416456 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16456048 Supplier Medchem Express Supplier No. HY14380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-3845 is a potent, selective, irreversible and orally active inhibitor of fatty acid amide hydrolase (FAAH), with a Ki of 0.23 μM. PF-3845 is a covalent inhibitor that carbamylates FAAH's serine nucleophile. PF-3845 can reduce pain sensation, inflammation, and anxiety/depression without substantial effects on motility or cognition.

PF-3845 is a potent, selective, irreversible and orally active inhibitor of fatty acid amide hydrolase (FAAH), with a Ki of 0.23 μM. PF-3845 is a covalent inhibitor that carbamylates FAAH's serine nucleophile. PF-3845 can reduce pain sensation, inflammation, and anxiety/depression without substantial effects on motility or cognition.

Specifications

Chemical Name or Material PF-3845
CAS 1196109-52-0
Color White
Purity Grade Notes Research
Molecular Formula C24H23F3N4O2
Quantity 10 mg
Solubility Information DMSO : ≥ 100 mg/mL (219.08 mM)
SMILES O=C(N1CCC(CC2=CC=CC(OC3=NC=C(C=C3)C(F)(F)F)=C2)CC1)NC4=CN=CC=C4
Molecular Weight (g/mol) 456.46
Formula Weight 456.46
Percent Purity 99.8%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.