missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 3084014 hydrobromide

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
PF 3084014 hydrobromide is a potent γ-secretase inhibitor (IC50 values are 1.2 and 6.2 nM in whole cell and cell-free assays, respectively). Reduces Aβ in brain, CSF and plasma in mice and guinea pigs.
Specifications
Specifications
| Chemical Name or Material | (2 S)-2-[[(2 S)-6,8-Difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]amino]-N-[1-[2-[(2,2-dimethylpropyl)amino]-1,1-dimethylethyl]-1 H-imidazol-4-yl]pentanamide dihydrobromide |
| CAS | 1962925-29-6 |
| Quantity | 10 mg |
| Target | gamma-Secretase |
| Molecular Formula | C27H41F2N5O.HBr |
| Formula Weight | 651.47 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction