missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 3084014 hydrobromide
GREENER_CHOICE

Product Code. 18758688
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18758688

Brand: Tocris Bioscience™ 5751/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent γ-secretase inhibitor

PF 3084014 hydrobromide is a potent γ-secretase inhibitor (IC50 values are 1.2 and 6.2 nM in whole cell and cell-free assays, respectively). Reduces Aβ in brain, CSF and plasma in mice and guinea pigs.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2 S)-2-[[(2 S)-6,8-Difluoro-1,2,3,4-tetrahydro-2-naphthalenyl]amino]-N-[1-[2-[(2,2-dimethylpropyl)amino]-1,1-dimethylethyl]-1 H-imidazol-4-yl]pentanamide dihydrobromide
CAS 1962925-29-6
Quantity 10 mg
Target gamma-Secretase
Molecular Formula C27H41F2N5O.HBr
Formula Weight 651.47
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.