missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PF-06751979, MedChemExpress
Description
PF-06751979 is a potent, brain penetrant, β-site amyloid precursor protein cleaving enzyme 1 (BACE1) inhibitor with an IC50 of 7.3 nM in BACE1 binding assay.
Specifications
Specifications
| Chemical Name or Material | PF-06751979 |
| CAS | 1818339-66-0 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C18H19F2N5O3S2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 150 mg/mL (329.31 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (109.77 mM; Need ultrasonic) |
| SMILES | FC(F)OC1=CN=C(C(NC2=CSC([C@]34CO[C@@H](C)C[C@@]3([H])CSC(N)=N4)=N2)=O)C=C1 |
| Molecular Weight (g/mol) | 455.5 |
| Formula Weight | 455.5 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?