missing translation for 'onlineSavingsMsg'
Learn More

PF-06726304, MedChemExpress

Product Code. 16430257
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16430257

Brand: Medchem Express HY103682

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PF-06726304 is a potent and selective EZH2 inhibitor. PF-06726304 inhibits wild-type and Y641N mutant EZH2 with Kis of 0.7 and 3.0 nM, respectively. PF-06726304 displays robust antitumor growth activity.

PF-06726304 is a potent and selective EZH2 inhibitor. PF-06726304 inhibits wild-type and Y641N mutant EZH2 with Kis of 0.7 and 3.0 nM, respectively. PF-06726304 displays robust antitumor growth activity.

Specifications

Chemical Name or Material PF-06726304
CAS 1616287-82-1
Color White
Purity Grade Notes Research
Molecular Formula C22H21Cl2N3O3
Quantity 50 mg
Solubility Information DMSO : ≥ 15 mg/mL (33.61 mM)
SMILES O=C1N(CC2=C(C)C=C(C)NC2=O)CCC3=C1C(Cl)=C(C4=C(C)ON=C4C)C=C3Cl
Molecular Weight (g/mol) 446.33
Formula Weight 446.33
Percent Purity 99.64%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.