missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PF 06726304 acetate

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
PF 06726304 acetate is a highly potent and SAM-competitive EZH2 lysine methyltransferase inhibitor (IC50 = 0.7 nM). Inhibits proliferation of Karpas-422 cells (non-Hodgkin's lymphoma) in vitro, and halts tumor growth and reduces intratumoral H3K27Me3 levels in mice.
Specifications
Specifications
| Chemical Name or Material | PF 06726304 acetate |
| Synonym | 5,8-Dichloro-2-[(1,2-dihydro-4,6-dimethyl-2-oxo-3-pyridinyl)methyl]-7-(3,5-dimethyl-4-isoxazolyl)-3,4-dihydro-1(2H)-isoquinolinone acetate |
| Quantity | 10 mg |
| Solubility | Soluble to 100mM in DMSO and to 100mM in ethanol |
| Molecular Formula | C22H21Cl2N3O3.C2H4O2 |
| Formula Weight | 506.38 |
| Physical Form | Solid |
| Color | White |
| Product Type | Bioactive Small Molecule |
| Purity | >97% |
For Research Use Only.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction