missing translation for 'onlineSavingsMsg'
Learn More

PF-06471553, MedChemExpress

Product Code. 16455146
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455146 100 mg 100mg
16444188 10 mg 10mg
16440537 50 mg 50mg
16432986 25 mg 25mg
16426907 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16455146 Supplier Medchem Express Supplier No. HY108339
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress PF-06471553 is a potent, selective and orally available monoacylglycerol acyltransferase 3 (MGAT3) inhibitor, with an IC50 of 92 nM.

PF-06471553 is a potent, selective and orally available monoacylglycerol acyltransferase 3 (MGAT3) inhibitor, with an IC50 of 92 nM.

Specifications

Chemical Name or Material PF-06471553
CAS 1808094-07-6
Color White
Purity Grade Notes Research
Molecular Formula C23H25N5O4S
Quantity 100 mg
Solubility Information DMSO : ≥ 150 mg/mL (320.83 mM)
SMILES O=S(C1=CC2=C(CN(C(CC3=CC=CC(OC)=C3)=O)C2)C=C1)(NC4=NN(C5CCC5)N=C4)=O
Molecular Weight (g/mol) 467.54
Formula Weight 467.54
Percent Purity 98.18%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.